Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Acetylacetophenone, MP Biomedicals

Catalog No. ICN203884 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN203884 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN203884 Supplier MP Biomedicals, Inc Supplier No. 0520388410

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1009-61-6
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00008750
InChI Key SKBBQSLSGRSQAJ-UHFFFAOYSA-N
Synonym 1,4-diacetylbenzene, 1,1'-1,4-phenylene diethanone, p-diacetylbenzene, p-acetylacetophenone, benzene, p-diacetyl, ethanone, 1,1'-1,4-phenylene bis, 1-4-acetylphenyl ethanone, 1-4-acetylphenyl ethan-1-one, 1,1-1,4-phenylene bis-ethanone, 4-acetylacetophenone
PubChem CID 13888
IUPAC Name 1-(4-acetylphenyl)ethan-1-one
SMILES CC(=O)C1=CC=C(C=C1)C(C)=O

Specifications

Quantity 10 g
Formula Weight 162.19
Chemical Name or Material p-Acetyl Acetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.