accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3C(S)ONHNHOBrCH3CH3NHO

CAS RN 100900-11-6

IUPAC Name: 2-bromo-N-(3-{[(3S)-1-hydroxy-2,2,5,5-tetramethylpyrrolidin-3-yl]formamido}propyl)acetamide
Synonyms: 2-bromo-N-(3-{[(3S)-1-hydroxy-2,2,5,5-tetramethylpyrrolidin-3-yl]formamido}propyl)acetamide
Molecular Weight (g/mol): 364.28
Molecular Formula: C14H26BrN3O3
InChi Key: SLPTVCQENYQDNA-SNVBAGLBSA-N
SMILES: CC1(C)C[C@H](C(=O)NCCCNC(=O)CBr)C(C)(C)N1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results