Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-[3-(2-Bromoacetamido)propylcarbamoyl]-PROXYL, Free Radical, Thermo Scientific™

Catalog No. AC224830250
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC224830250 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC224830250 Supplier Thermo Scientific Chemicals Supplier No. 224830250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100900-11-6
Molecular Formula C14H25BrN3O3
Molecular Weight (g/mol) 363.276
InChI Key HRHTYNXIXOOARI-UHFFFAOYSA-N
Synonym n-3-2-bromoacetyl amino propyl-1, 3-3-2-bromoacetamido propylcarbamoyl-proxyl free radical
PubChem CID 2724300
IUPAC Name N-[3-[(2-bromoacetyl)amino]propyl]-1-$l^{1}-oxidanyl-2,2,5,5-tetramethylpyrrolidine-3-carboxamide
SMILES CC1(CC(C(N1[O])(C)C)C(=O)NCCCNC(=O)CBr)C

Specifications

Quantity 25 mg
Formula Weight 363.26
Chemical Name or Material 3-[3-(2-Bromoacetamido)propylcarbamoyl]-PROXYL
Hazard Category LIGHT SENSITIVE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.