accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1009582-56-2

IUPAC Name: 2,4-dibromo-6-[({[1-(4-methylbenzenesulfonyl)-4,5-diphenyl-4,5-dihydro-1H-imidazol-2-yl]methyl}(1-phenylethyl)amino)methyl]phenol
Synonyms: 2,4-dibromo-6-[({[1-(4-methylbenzenesulfonyl)-4,5-diphenyl-4,5-dihydroimidazol-2-yl]methyl}(1-phenylethyl)amino)methyl]phenol
Molecular Weight (g/mol): 773.58
Molecular Formula: C38H35Br2N3O3S
InChi Key: QMNHSXCNPYPSMG-UHFFFAOYNA-N
SMILES: CC(N(CC1=NC(C(N1S(=O)(=O)C1=CC=C(C)C=C1)C1=CC=CC=C1)C1=CC=CC=C1)CC1=CC(Br)=CC(Br)=C1O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results