Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IAP 90.0+%, TCI America™
SDP

Catalog No. D416850MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D416850MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D416850MG Supplier TCI America Supplier No. D416850MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1009582-56-2
Molecular Formula C38H35Br2N3O3S
Molecular Weight (g/mol) 773.584
InChI Key QMNHSXCNPYPSMG-GILITBBJSA-N
Synonym IAP
PubChem CID 24822536
IUPAC Name 2,4-dibromo-6-[[[(4S,5S)-1-(4-methylphenyl)sulfonyl-4,5-diphenyl-4,5-dihydroimidazol-2-yl]methyl-[(1S)-1-phenylethyl]amino]methyl]phenol
SMILES CC1=CC=C(C=C1)S(=O)(=O)N2C(C(N=C2CN(CC3=CC(=CC(=C3O)Br)Br)C(C)C4=CC=CC=C4)C5=CC=CC=C5)C6=CC=CC=C6

Specifications

Color Yellow
Quantity 50 mg
Formula Weight 773.58
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dibromo-6-[[[[(4S,5S)-4,5-dihydro-4,5-diphenyl-1-tosyl-1H-imidazol-2-yl]methyl][(S)-1-phenylethy
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.