accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 100960-07-4

HNNNH2

CAS RN 100960-07-4

IUPAC Name: 1H-pyrrolo[2,3-b]pyridin-5-amine
Synonyms: 1H-pyrrolo[2,3-b]pyridin-5-amine
Molecular Weight (g/mol): 133.15
Molecular Formula: C7H7N3
InChi Key: PLWBENCHEUFMTN-UHFFFAOYSA-N
SMILES: NC1=CN=C2NC=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results