accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10097-28-6

IUPAC Name: oxidaniumylidynesilanide
Synonyms: silicon monoxide SiO
Molecular Weight (g/mol): 44.08
Molecular Formula: OSi
InChi Key: LIVNPJMFVYWSIS-UHFFFAOYSA-N
SMILES: [O+]#[Si-]
Molecular Weight (g/mol) 
  • (3)
  • (10)
Quantity 
  • (1)
  • (4)
  • (1)
  • (3)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (4)
  • (1)
Physical Form 
  • (2)
  • (3)
  • (2)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (3)
  • (10)
  • (3)
  • (10)
  • (1)
  • (4)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (2)
  • (4)