Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silicon(II) oxide, 99.99% (metals basis)

Catalog No. AA3634722
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3634722 100 g
AA3634714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3634722 Supplier Thermo Scientific Chemicals Supplier No. 03634722
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 10097-28-6 | OSi | 44.084 g/mol

A major application of silicon oxide is production of portland cement. It is used in food and pharmaceutical applications and in semiconductor industry. It is the major precursor for preparing glass and silicon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10097-28-6
Molecular Formula OSi
Molecular Weight (g/mol) 44.084
MDL Number MFCD00151536
InChI Key LIVNPJMFVYWSIS-UHFFFAOYSA-N
Synonym silicon monoxide, silylene, oxo, silicon ii oxide, unii-1oqn9cbg7l, 1oqn9cbg7l, silicon ii oxide, optical grade, sio, oxidosilicon, oxosilanylidene, oxosilylene
PubChem CID 66241
ChEBI CHEBI:30588
IUPAC Name oxoniumylidynesilanide
SMILES [O+]#[Si-]

Specifications

Boiling Point 1880°C
Melting Point >1702°C
Color Black
Physical Form Lump
Quantity 100 g
Odor Odorless
Assay Percent Range 99.99% (metals basis)
Density 2.13
Solubility Information Insoluble in water.
Formula Weight 44.09
Chemical Name or Material Silicon(II) oxide
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 233-232-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.