accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 101-42-8

H3CNCH3ONH

CAS RN 101-42-8

IUPAC Name: 3,3-dimethyl-1-phenylurea
Synonyms: 1,1-dimethyl-3-phenylurea
Molecular Weight (g/mol): 164.21
Molecular Formula: C9H12N2O
InChi Key: XXOYNJXVWVNOOJ-UHFFFAOYSA-N
SMILES: CN(C)C(=O)NC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results