accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3

CAS RN 101-48-4

IUPAC Name: (2,2-dimethoxyethyl)benzene
Synonyms: benzene, (2,2-dimethoxyethyl)-
Molecular Weight (g/mol): 166.22
Molecular Formula: C10H14O2
InChi Key: WNJSKZBEWNVKGU-UHFFFAOYSA-N
SMILES: COC(CC1=CC=CC=C1)OC
Boiling Point 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)