accessibility menu, dialog, popup

UserName

Viewing profile as user:
OH

CAS RN 101-53-1

IUPAC Name: 4-benzylphenol
Synonyms: benzophenol 4-hydroxydiphenylmethane
Molecular Weight (g/mol): 184.24
Molecular Formula: C13H12O
InChi Key: HJSPWKGEPDZNLK-UHFFFAOYSA-N
SMILES: OC1=CC=C(CC2=CC=CC=C2)C=C1
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)