Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzylphenol, Spectrum™ Chemical
SDP

Catalog No. 18620860 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-860 100 g
18-620-859 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-620-860 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. H2038100GM
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synonym: p-Benzylphenol, 4-Hydroxydiphenylmethane, Fesiasept

Chemical Identifiers

CAS 101-53-1
Molecular Formula C13H12O
Molecular Weight (g/mol) 184.24
InChI Key HJSPWKGEPDZNLK-UHFFFAOYSA-N
IUPAC Name 4-benzylphenol
SMILES OC1=CC=C(CC2=CC=CC=C2)C=C1

Specifications

Boiling Point 330°C
Packaging Amber Bottle
Quantity 100 g
Percent Purity 98%
Physical Form Crystals or Flakes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.