accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 101-92-8

IUPAC Name: N-(4-chlorophenyl)-3-oxobutanamide
Synonyms: 4'-chloroacetoacetanilide p-chlorodiacetanilide acetoacet-p-chloroanilide acetoacetyl-4-chloroanilide C10H10ClNO2
Molecular Weight (g/mol): 211.65
Molecular Formula: C10H10ClNO2
InChi Key: JMRJWEJJUKUBEA-UHFFFAOYSA-N
SMILES: CC(=O)CC(=O)NC1=CC=C(Cl)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results