accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 101-96-2

IUPAC Name: N1,N4-bis(butan-2-yl)benzene-1,4-diamine
Synonyms: N1,N4-bis(sec-butyl)benzene-1,4-diamine
Molecular Weight (g/mol): 220.36
Molecular Formula: C14H24N2
InChi Key: FSWDLYNGJBGFJH-UHFFFAOYNA-N
SMILES: CCC(C)NC1=CC=C(NC(C)CC)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)