Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Di-sec-butyl-1,4-phenylenediamine 98.0+%, TCI America™
SDP

Catalog No. D226825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D226825ML 25 mL
D2268500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D226825ML Supplier TCI America Supplier No. D226825ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 101-96-2
Molecular Formula C14H24N2
Molecular Weight (g/mol) 220.36
MDL Number MFCD00043658
InChI Key FSWDLYNGJBGFJH-UHFFFAOYSA-N
PubChem CID 7589
IUPAC Name 1-N,4-N-di(butan-2-yl)benzene-1,4-diamine
SMILES CCC(C)NC1=CC=C(C=C1)NC(C)CC

Specifications

Boiling Point 159°C
Quantity 25 mL
UN Number 2922
Formula Weight 220.36
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N′-Di-sec-butyl-1,4-phenylenediamine
EINECSNumber (3)-0247
RTECSNumber SS9040000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.