accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 101-98-4

IUPAC Name: benzyl(2-hydroxyethyl)methylazanium
Synonyms: benzyl(2-hydroxyethyl)methylazanium
Molecular Weight (g/mol): 166.24
Molecular Formula: C10H16NO
InChi Key: WOUANPHGFPAJCA-UHFFFAOYNA-O
SMILES: C[NH+](CCO)CC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results