CAS RN 1010-60-2
IUPAC Name:
2-chloro-1,4-dihydronaphthalene-1,4-dione
Synonyms:
2-chloro-1,4-naphthoquinone C10H5ClO2
Molecular Weight (g/mol):
192.6
Molecular Formula:
C10H5ClO2
InChi Key:
CCTJHVLTAJTPBV-UHFFFAOYSA-N
SMILES:
ClC1=CC(=O)C2=CC=CC=C2C1=O