Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-1,4-naphthoquinone 98.0+%, TCI America™
SDP

Catalog No. C27215G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C27215G Supplier TCI America Supplier No. C27215G

Specifications

CAS 1010-60-2
Melting Point 116°C
Color Yellow
Molecular Formula C10H5ClO2
Quantity 5 g
MDL Number MFCD00029187
InChI Key CCTJHVLTAJTPBV-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C(=O)C=C(C2=O)Cl
IUPAC Name 2-chloronaphthalene-1,4-dione
Molecular Weight (g/mol) 192.598
PubChem CID 13891
ChEBI CHEBI:28160
Formula Weight 192.60
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-1,4-naphthoquinone
Show More Show Less
RTECSNumber QL7450000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.