accessibility menu, dialog, popup

UserName

Viewing profile as user:
ZrOSiOOO

CAS RN 10101-52-7

IUPAC Name: zirconium(4+) silicate
Synonyms: zirconium(IV) silicate
Molecular Weight (g/mol): 183.31
Molecular Formula: O4SiZr
InChi Key: GFQYVLUOOAAOGM-UHFFFAOYSA-N
SMILES: [Zr+4].[O-][Si]([O-])([O-])[O-]
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)