Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zirconium Silicate, 99.5%, MP Biomedicals

Catalog No. ICN220011 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN220011 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN220011 Supplier MP Biomedicals, Inc Supplier No. 0522001180

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10101-52-7
Molecular Formula O4SiZr
Molecular Weight (g/mol) 183.31
MDL Number MFCD00085353
InChI Key GFQYVLUOOAAOGM-UHFFFAOYSA-N
Synonym zirconium silicate, hyacinth, zircon, zircon zr sio4, zirconite, zircosil 15, standard sf 200, ultrox 500w, a-pax 45m, zirconium iv silicate
PubChem CID 61775
IUPAC Name zirconium(4+) silicate
SMILES [Zr+4].[O-][Si]([O-])([O-])[O-]

Specifications

Quantity 100 g
Formula Weight 183.3
Chemical Name or Material Zirconium Silicate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.