accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1010120-55-4

IUPAC Name: 5-bromo-[1,2,4]triazolo[1,5-a]pyridin-2-amine
Synonyms: 5-bromo-[1,2,4]triazolo[1,5-a]pyridin-2-amine
Molecular Weight (g/mol): 213.04
Molecular Formula: C6H5BrN4
InChi Key: TVGFHUIJNZRKFW-UHFFFAOYSA-N
SMILES: NC1=NN2C(C=CC=C2Br)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results