accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClH3CNCH3CH3H3C

CAS RN 10108-87-9

IUPAC Name: decyltrimethylazanium chloride
Synonyms: decyltrimethylammonium chloride
Molecular Weight (g/mol): 235.84
Molecular Formula: C13H30ClN
InChi Key: HXWGXXDEYMNGCT-UHFFFAOYSA-M
SMILES: [Cl-].CCCCCCCCCC[N+](C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results