accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 101084-81-5

IUPAC Name: 3-(dihydroxyboranyl)-5-nitrobenzoic acid
Synonyms: 3-(dihydroxyboranyl)-5-nitrobenzoic acid
Molecular Weight (g/mol): 210.94
Molecular Formula: C7H6BNO6
InChi Key: WNIFCLWDGNHGMX-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(=CC(=C1)[N+]([O-])=O)C(O)=O
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results