Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Carboxy-5-nitrophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. C23855G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2385-5G 5 g
C23851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2385-5G Supplier TCI America Supplier No. C23855G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 101084-81-5
Melting Point 229°C
Color White-Yellow
Molecular Formula C7H6BNO6
Quantity 5 g
MDL Number MFCD00757433
Synonym 3-carboxy-5-nitrophenylboronic acid, 3-nitro-5-carboxyphenylboronic acid, 5-nitro-3-carboxyphenylboronic acid, 3-carboxy-5-nitrophenyl boronic acid, 3-carboxy-5-nitrobenzeneboronic acid
InChI Key WNIFCLWDGNHGMX-UHFFFAOYSA-N
SMILES B(C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)O)(O)O
IUPAC Name 3-borono-5-nitrobenzoic acid
Molecular Weight (g/mol) 210.936
PubChem CID 2773315
Formula Weight 210.94
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.