accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COHH3CH3CCH3CH3CH3OHCH3H3CCH3(?)

CAS RN 101203-31-0

IUPAC Name: 3,3'-di-tert-butyl-5,5',6,6'-tetramethyl-[1,1'-biphenyl]-2,2'-diol
Synonyms: 3,3'-di-tert-butyl-5,5',6,6'-tetramethyl-[1,1'-biphenyl]-2,2'-diol
Molecular Weight (g/mol): 354.53
Molecular Formula: C24H34O2
InChi Key: NMVVBVMYPLMIOU-UHFFFAOYSA-N
SMILES: CC1=CC(=C(O)C(=C1C)C1=C(C)C(C)=CC(=C1O)C(C)(C)C)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results