CAS RN 10124-43-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Molecular Sieve Deperox/Dehydrate, Solstice
Beads, for the deperoxidation and drying of solvents