accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2ClHg

CAS RN 10124-48-8

IUPAC Name: mercury(2+) azanide chloride
Synonyms: mercuric amidochloride
Molecular Weight (g/mol): 252.06
Molecular Formula: ClH2HgN
InChi Key: WRWRKDRWMURIBI-UHFFFAOYSA-M
SMILES: [NH2-].[Cl-].[Hg++]
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (4)
Percent Purity 
  • (4)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (1)
  • (4)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (4)
  • (4)