accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNH2H3C

CAS RN 1013-76-9

IUPAC Name: 4-(2,4-dimethylphenyl)piperazin-1-ium
Synonyms: 4-(2,4-dimethylphenyl)piperazin-1-ium
Molecular Weight (g/mol): 191.3
Molecular Formula: C12H19N2
InChi Key: RUIMBVCRNZHCRQ-UHFFFAOYSA-O
SMILES: CC1=CC=C(N2CC[NH2+]CC2)C(C)=C1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)