Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2,4-Dimethylphenyl)piperazine 98.0+%, TCI America™
SDP

Catalog No. D41365G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D41365G Supplier TCI America Supplier No. D41365G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1013-76-9
Molecular Formula C12H18N2
Molecular Weight (g/mol) 190.29
MDL Number MFCD00023127
InChI Key RUIMBVCRNZHCRQ-UHFFFAOYSA-N
Synonym 1-2,4-dimethylphenyl piperazine, 1-2,4-xylyl piperazine, piperazine, 1-2,4-dimethylphenyl, 2,4-dimethylphenyl piperazine, 1-2,4-dimethylphenyl piperazin, pubchem8588, akos bb-5739, timtec-bb sbb003650, 4-dimethylphenyl piperazine, acmc-2097uz
PubChem CID 70544
IUPAC Name 1-(2,4-dimethylphenyl)piperazine
SMILES CC1=CC(=C(C=C1)N2CCNCC2)C

Specifications

Color Yellow
Boiling Point 153°C
Quantity 5 g
UN Number 3267
Formula Weight 190.29
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-(2,4-Dimethylphenyl)piperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.