accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1013-96-3

IUPAC Name: (2E)-3-(2-nitrophenyl)prop-2-enoic acid
Synonyms: (2E)-3-(2-nitrophenyl)prop-2-enoic acid
Molecular Weight (g/mol): 193.16
Molecular Formula: C9H7NO4
InChi Key: BBQDLDVSEDAYAA-AATRIKPKSA-N
SMILES: OC(=O)\C=C\C1=CC=CC=C1[N+]([O-])=O
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)