Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Nitrocinnamic Acid 98.0+%, TCI America™
SDP

Catalog No. N035625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N035625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N035625G Supplier TCI America Supplier No. N035625G

missing translation for 'validMainframeMsg'

Specifications

CAS 1013-96-3
Color Yellow
Molecular Formula C9H7NO4
Quantity 25 g
MDL Number MFCD00007189
Synonym 2-nitrocinnamic acid, o-nitrocinnamic acid, 3-2-nitrophenyl acrylic acid, e-3-2-nitrophenyl acrylic acid, cinnamic acid, o-nitro, trans-2-nitrocinnamic acid, 2e-3-2-nitrophenyl prop-2-enoic acid, 2-propenoic acid, 3-2-nitrophenyl, 3-2-nitrophenyl-acrylic acid, ccris 1668
InChI Key BBQDLDVSEDAYAA-AATRIKPKSA-N
SMILES C1=CC=C(C(=C1)C=CC(=O)O)[N+](=O)[O-]
Molecular Weight (g/mol) 193.158
PubChem CID 735923
Formula Weight 193.16
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans-2-Nitrocinnamic Acid
Show More Show Less
RTECSNumber UD3642850
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.