accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10130-29-7

IUPAC Name: sodium 6-methyl-2-(4-{2-[2-oxo-1-(phenylcarbamoyl)propylidene]hydrazin-1-yl}phenyl)-1,3-benzothiazole-7-sulfonate
Synonyms: sodium 6-methyl-2-(4-{2-[2-oxo-1-(phenylcarbamoyl)propylidene]hydrazin-1-yl}phenyl)-1,3-benzothiazole-7-sulfonate
Molecular Weight (g/mol): 530.55
Molecular Formula: C24H19N4NaO5S2
InChi Key: OZRXNTRXLHTVPI-UHFFFAOYSA-M
SMILES: [Na+].CC(=O)C(=NNC1=CC=C(C=C1)C1=NC2=CC=C(C)C(=C2S1)S([O-])(=O)=O)C(=O)NC1=CC=CC=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results