accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OSClOO

CAS RN 10130-87-7

IUPAC Name: 2-methoxybenzene-1-sulfonyl chloride
Synonyms: 2-methoxybenzenesulfonyl chloride
Molecular Weight (g/mol): 206.64
Molecular Formula: C7H7ClO3S
InChi Key: GYOBZOBUOMDRRN-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)