Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxybenzenesulfonyl chloride, 95%

Catalog No. AAH34455MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH34455MD 250 mg
AAH3445503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH34455MD Supplier Thermo Scientific Chemicals Supplier No. H34455MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 10130-87-7 | C7H7ClO3S | 206.64 g/mol

For synthesis and pharmacological evaluation of sulfonamide derivatives of thiazolidin-4-ones 5-Bromo-2-methoxybenzenesulfonyl chloride is used.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
For synthesis and pharmacological evaluation of sulfonamide derivatives of thiazolidin-4-ones 5-Bromo-2-methoxybenzenesulfonyl chloride is used.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10130-87-7
Molecular Formula C7H7ClO3S
Molecular Weight (g/mol) 206.64
MDL Number MFCD01961367
InChI Key GYOBZOBUOMDRRN-UHFFFAOYSA-N
Synonym 2-methoxy-benzenesulfonyl chloride, 2-methoxybenzene-1-sulfonyl chloride, 2-methoxybenzenesulfonylchloride, o-anisolesulfonyl chloride, 2-methoxyphenylsulfonyl chloride, acmc-1bntp, methoxyphenylsulfonyl chloride, 2-methoxybenzensulfonylchloride, 2-methoxybenzenesulphonylchloride, 2-methoxybenzene sulfonylchloride
PubChem CID 3745851
IUPAC Name 2-methoxybenzenesulfonyl chloride
SMILES COC1=CC=CC=C1S(=O)(=O)Cl

Specifications

Melting Point 50°C to 54°C
Quantity 250 mg
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 206.65
Percent Purity 97%
Chemical Name or Material 2-Methoxybenzenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.