accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10130-89-9

IUPAC Name: 4-(chlorosulfonyl)benzoic acid
Synonyms: 4-(chlorosulfonyl)benzoic acid
Molecular Weight (g/mol): 220.62
Molecular Formula: C7H5ClO4S
InChi Key: PTCSSXYPZOFISK-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)S(Cl)(=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (6)
  • (3)
  • (5)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)