Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Chlorosulfonyl)benzoic acid, 96%

Catalog No. AC405060250
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC405060250 25 g Glass bottle
AC405060050 5 g Glass bottle
AC405061000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC405060250 Supplier Thermo Scientific Chemicals Supplier No. 405060250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 10130-89-9 | C7H5ClO4S | 220.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10130-89-9
Molecular Formula C7H5ClO4S
Molecular Weight (g/mol) 220.63
InChI Key PTCSSXYPZOFISK-UHFFFAOYSA-N
Synonym 4-chlorosulfonyl benzoic acid, benzoic acid, 4-chlorosulfonyl, p-chlorosulfonyl benzoic acid, 4-chlorosulphonyl benzoic acid, p-carboxybenzenesulfonyl chloride, benzoic acid, p-chlorosulfonyl, 4-chlorosulfonyl-benzoic acid, 4-carboxybenzenesulfonyl chloride, 4-carboxy-benzenesulfonylchloride, pubchem14811
PubChem CID 82385
IUPAC Name 4-chlorosulfonylbenzoic acid
SMILES C1=CC(=CC=C1C(=O)O)S(=O)(=O)Cl

Specifications

Melting Point 230.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula CO2HC6H4SO2Cl
Quantity 25 g
Solubility Information Solubility in water: decomposes.
Formula Weight 220.63
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 4-(Chlorosulfonyl)benzoic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 233-367-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.