accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2ClNClN

CAS RN 10132-07-7

IUPAC Name: 2,6-dichloropyrimidin-4-amine
Synonyms: 2,6-dichloropyrimidin-4-amine
Molecular Weight (g/mol): 163.99
Molecular Formula: C4H3Cl2N3
InChi Key: UPVBKNZVOJNQKE-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=NC(Cl)=N1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)