Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-2,6-dichloropyrimidine 98.0+%, TCI America™
SDP

Catalog No. A208225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A208225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A208225G Supplier TCI America Supplier No. A208225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10132-07-7
Molecular Formula C4H3Cl2N3
Molecular Weight (g/mol) 163.989
MDL Number MFCD00038015
InChI Key UPVBKNZVOJNQKE-UHFFFAOYSA-N
Synonym 4-amino-2,6-dichloropyrimidine, 6-amino-2,4-dichloropyrimidine, 4-pyrimidinamine, 2,6-dichloro, 2,4-dichloro-6-aminopyrimidine, 2,6-dichloro-4-aminopyrimidine, 2,4-dichloro-6-amino pyrimidine, 2,6-dichloro-pyrimidin-4-ylamine, 2,6-dichloropyrimidine-4-ylamine, 2,6-dichlorpyrimidin-4-amin, pyrimidine, 4-amino-2,6-dichloro
PubChem CID 82387
IUPAC Name 2,6-dichloropyrimidin-4-amine
SMILES C1=C(N=C(N=C1Cl)Cl)N

Specifications

Melting Point 254°C
Color Yellow
Quantity 25 g
Formula Weight 163.99
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2,6-dichloropyrimidine
EINECSNumber (9)-0074&(9)-0588
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.