accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10138-41-7

IUPAC Name: erbium(3+) trichloride
Synonyms: erbium(III) chloride
Molecular Weight (g/mol): 273.61
Molecular Formula: Cl3Er
InChi Key: HDGGAKOVUDZYES-UHFFFAOYSA-K
SMILES: [Cl-].[Cl-].[Cl-].[Er+3]
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (4)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (4)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)