accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHN(R)(R)OH(S)OH(R)HOO(R)O

CAS RN 10139-04-5

IUPAC Name: N-[(2R,3R,4R,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-phenoxyoxan-3-yl]acetamide
Synonyms: N-[(2R,3R,4R,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-phenoxyoxan-3-yl]acetamide
Molecular Weight (g/mol): 297.31
Molecular Formula: C14H19NO6
InChi Key: ZUJDLWWYFIZERS-KSTCHIGDSA-N
SMILES: CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@@H]1OC1=CC=CC=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results