Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl N-Acetyl-alpha-D-glucosaminide, TCI America™
SDP

Catalog No. P0130100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0130100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P0130100MG Supplier TCI America Supplier No. P0130100MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10139-04-5
Molecular Formula C14H19NO6
Molecular Weight (g/mol) 297.307
MDL Number MFCD00067651
InChI Key ZUJDLWWYFIZERS-PDWCTOEPSA-N
Synonym Phenyl 2-Acetamido-2-deoxy-alpha-D-glucopyranoside
PubChem CID 133655432
IUPAC Name N-[(2S,3S,4S,5S,6S)-4,5-dihydroxy-6-(hydroxymethyl)-2-phenoxyoxan-3-yl]acetamide
SMILES CC(=O)NC1C(C(C(OC1OC2=CC=CC=C2)CO)O)O

Specifications

Melting Point 241°C
Color White
Quantity 100 mg
Formula Weight 297.31
Physical Form Crystalline Powder
Chemical Name or Material Phenyl N-Acetyl-alpha-D-glucosaminide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.