accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10147-71-4

IUPAC Name: 2-chloro-N-(3-nitrophenyl)acetamide
Synonyms: 2-chloro-N-(3-nitrophenyl)acetamide
Molecular Weight (g/mol): 214.61
Molecular Formula: C8H7ClN2O3
InChi Key: UCBHRCPNMDOUMV-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC(NC(=O)CCl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results