accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CBr

CAS RN 101532-38-1

IUPAC Name: 1-bromo-4-(4-heptylcyclohexyl)benzene
Synonyms: 1-bromo-4-(4-heptylcyclohexyl)benzene
Molecular Weight (g/mol): 337.35
Molecular Formula: C19H29Br
InChi Key: OKIVMWBXCXBOGL-UHFFFAOYSA-N
SMILES: CCCCCCCC1CCC(CC1)C1=CC=C(Br)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results