accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10154-75-3

IUPAC Name: 3-(benzenesulfonyl)propanenitrile
Synonyms: 3-(benzenesulfonyl)propanenitrile
Molecular Weight (g/mol): 195.24
Molecular Formula: C9H9NO2S
InChi Key: OVZPZPVSUAKTCM-UHFFFAOYSA-N
SMILES: O=S(=O)(CCC#N)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results