accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONOCH3OCH3Cl

CAS RN 1016825-43-6

IUPAC Name: ethyl 4-chloro-6,8-dimethoxyquinoline-3-carboxylate
Synonyms: ethyl 4-chloro-6,8-dimethoxyquinoline-3-carboxylate
Molecular Weight (g/mol): 295.72
Molecular Formula: C14H14ClNO4
InChi Key: DSGVVCRDDXQCTD-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN=C2C(OC)=CC(OC)=CC2=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results