accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2COOOSOO

CAS RN 1017587-57-3

IUPAC Name: 3,5-bis(benzyloxy)-2-[(benzyloxy)methyl]-6-(phenylsulfanyl)-4-(prop-2-en-1-yloxy)oxane
Synonyms: 3,5-bis(benzyloxy)-2-[(benzyloxy)methyl]-6-(phenylsulfanyl)-4-(prop-2-en-1-yloxy)oxane
Molecular Weight (g/mol): 582.76
Molecular Formula: C36H38O5S
InChi Key: OZARVANREPUDBF-UHFFFAOYNA-N
SMILES: C=CCOC1C(OCC2=CC=CC=C2)C(COCC2=CC=CC=C2)OC(SC2=CC=CC=C2)C1OCC1=CC=CC=C1
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)