Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl 3-O-Allyl-2,4,6-tri-O-benzyl-1-thio-beta-D-galactopyranoside 98.0+%, TCI America™
SDP

Catalog No. P16601G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P16601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P16601G Supplier TCI America Supplier No. P16601G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity thiogalactoside derivative for use as a laboratory reagent.

Chemical Identifiers

CAS 1017587-57-3
Molecular Formula C36H38O5S
Molecular Weight (g/mol) 582.755
InChI Key OZARVANREPUDBF-JZPVOVDPSA-N
Synonym Gal[246Bn,3All]-beta-SPh
PubChem CID 133554193
IUPAC Name (2R,3R,4R,5R,6R)-3,5-bis(phenylmethoxy)-2-(phenylmethoxymethyl)-6-phenylsulfanyl-4-prop-2-enoxyoxane
SMILES C=CCOC1C(C(OC(C1OCC2=CC=CC=C2)SC3=CC=CC=C3)COCC4=CC=CC=C4)OCC5=CC=CC=C5

Specifications

Color White
Quantity 1 g
Formula Weight 582.76
Percent Purity ≥98.0% (HPLC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material Phenyl 3-O-Allyl-2,4,6-tri-O-benzyl-1-thio-beta-D-galactopyranoside
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.