accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OFOHF

CAS RN 1017779-45-1

IUPAC Name: 4-ethoxy-3,5-difluorophenol
Synonyms: 4-ethoxy-3,5-difluorophenol
Molecular Weight (g/mol): 174.15
Molecular Formula: C8H8F2O2
InChi Key: MRARGBDWPXWTCL-UHFFFAOYSA-N
SMILES: CCOC1=C(F)C=C(O)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results