accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3CNH3CCH3H3C

CAS RN 101827-46-7

IUPAC Name: hydrogen [(4-tert-butylphenyl)methyl](methyl)[(naphthalen-1-yl)methyl]amine chloride
Synonyms: hydrogen butenafine chloride
Molecular Weight (g/mol): 353.93
Molecular Formula: C23H28ClN
InChi Key: LJBSAUIFGPSHCN-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN(CC1=CC=C(C=C1)C(C)(C)C)CC1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results