accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10185-69-0

IUPAC Name: 3-(5-methyl-1,2,4-oxadiazol-3-yl)aniline
Synonyms: 3-(5-methyl-1,2,4-oxadiazol-3-yl)aniline
Molecular Weight (g/mol): 175.19
Molecular Formula: C9H9N3O
InChi Key: CTRGRIHPFAVSOF-UHFFFAOYSA-N
SMILES: CC1=NC(=NO1)C1=CC(N)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results